The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002212.3 | NP_002203.1 | 1 - 2 | 94 | 66 |
NM_181466.2 | NP_852131.1 | 1 - 2 | 94 | 66 | |
NR_052022.1 | 1 - 2 | 94 | 66 | ||
NR_052023.1 | 1 - 2 | 94 | 66 | ||
GenBank mRNA | AK296076.1 | - | 1 - 2 | 40 | 66 |
AK314969.1 | - | 1 - 2 | 70 | 66 | |
BC001119.2 | - | 1 - 2 | 67 | 66 | |
BC011845.2 | - | 1 - 2 | 45 | 66 | |
BC095465.1 | - | 1 - 2 | 43 | 66 | |
BG334381.1 | - | - | 48 | 66 | |
CT002623.1 | - | 1 - 2 | 46 | 66 |