The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207048.1 | NP_001193977.1 | 5 - 6 | 772 | 72 |
NM_001207049.1 | NP_001193978.1 | 5 - 6 | 697 | 72 | |
NM_012104.4 | NP_036236.1 | 6 - 7 | 1410 | 72 | |
NM_138971.3 | NP_620427.1 | 6 - 7 | 1278 | 72 | |
NM_138972.3 | NP_620428.1 | 6 - 7 | 1335 | 72 | |
NM_138973.3 | NP_620429.1 | 6 - 7 | 1203 | 72 | |
GenBank mRNA | AB032975.2 | - | 6 - 7 | 1392 | 72 |
AB050436.1 | - | 6 - 7 | 877 | 72 | |
AB050437.1 | - | 6 - 7 | 820 | 72 | |
AB050438.1 | - | 6 - 7 | 745 | 72 | |
AB177847.1 | - | 5 - 6 | 634 | 72 | |
AF161367.1 | - | 2 - 3 | 238 | 72 | |
AF190725.1 | - | 6 - 7 | 1402 | 72 | |
AF200193.1 | - | 6 - 7 | 910 | 72 | |
AF200343.1 | - | 6 - 7 | 949 | 72 | |
AF201468.1 | - | 6 - 7 | 1403 | 72 | |
AF204943.1 | - | 6 - 7 | 949 | 72 | |
AF338816.1 | - | 6 - 7 | 874 | 72 | |
AF338817.1 | - | 6 - 7 | 817 | 72 | |
AK075049.1 | - | 6 - 7 | 885 | 72 | |
AK296015.1 | - | 5 - 6 | 600 | 72 | |
AK296535.1 | - | 5 - 6 | 685 | 72 | |
BC036084.1 | - | 6 - 7 | 1405 | 72 | |
BC065492.1 | - | 6 - 7 | 1410 | 72 |