The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017832.3 | NP_060302.1 | 2 - 3 | 217 | 80 |
XM_011518816.2 | XP_011517118.1 | 2 - 3 | 213 | 80 | |
GenBank mRNA | AF153417.1 | - | 2 - 3 | 147 | 80 |
AK000464.1 | - | 2 - 3 | 201 | 80 | |
BC015795.1 | - | 2 - 3 | 429 | 80 |