The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014453.3 | NP_055268.1 | 4 - 5 | 644 | 67 |
NM_198426.2 | NP_940818.1 | 4 - 5 | 646 | 67 | |
XM_005258746.2 | XP_005258803.1 | 4 - 5 | 1144 | 67 | |
XM_005258747.3 | XP_005258804.1 | 4 - 5 | 1267 | 67 | |
GenBank mRNA | AF042384.1 | - | 4 - 5 | 612 | 67 |
AK311974.1 | - | 4 - 5 | 532 | 67 | |
AY364248.1 | - | 3 - 4 | 483 | 67 | |
BC002502.2 | - | 4 - 5 | 616 | 67 | |
BE856471.1 | - | 2 - 3 | 67 | ||
BT007298.1 | - | 3 - 4 | 483 | 67 | |
BU147101.1 | - | 2 - 3 | 291 | 67 | |
CN305066.1 | - | 4 - 5 | 647 | 67 | |
CR457002.1 | - | 3 - 4 | 483 | 67 |