The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021196.3 | NP_067019.3 | 18 - 19 | 2602 | 64 |
NM_133478.2 | NP_597812.1 | 23 - 24 | 2998 | 64 | |
GenBank mRNA | AF243499.2 | - | 18 - 19 | 2600 | 64 |
AF293337.2 | - | 22 - 23 | 2947 | 64 | |
AF293338.2 | - | 18 - 19 | 2600 | 64 | |
AF452248.1 | - | 20 - 21 | 2781 | 64 | |
BC109221.2 | - | 17 - 18 | 2343 | 64 |