The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142928.1 | NP_001136400.1 | 1 - 2 | 355 | 84 |
GenBank mRNA | AK055954.1 | - | 1 - 2 | 271 | 84 |
DB001467.1 | - | 1 - 2 | 365 | 84 |