The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143783.1 | NP_001137255.1 | 11 - 12 | 1534 | 80 |
NM_001143784.1 | NP_001137256.1 | 11 - 12 | 1498 | 80 | |
NM_001143785.1 | NP_001137257.1 | 11 - 12 | 1443 | 80 | |
NM_002005.3 | NP_001996.1 | 13 - 14 | 1804 | 80 | |
XM_005254880.1 | XP_005254937.1 | 12 - 13 | 1619 | 80 | |
XM_005254882.1 | XP_005254939.1 | 12 - 13 | 1610 | 80 | |
XM_017022005.1 | XP_016877494.1 | 13 - 14 | 1788 | 80 | |
XM_017022006.1 | XP_016877495.1 | 12 - 13 | 1661 | 80 | |
XM_017022007.1 | XP_016877496.1 | 12 - 13 | 1578 | 80 | |
XM_017022008.1 | XP_016877497.1 | 11 - 12 | 1452 | 80 | |
XM_017022009.1 | XP_016877498.1 | 13 - 14 | 1885 | 80 | |
XM_017022010.1 | XP_016877499.1 | 12 - 13 | 1736 | 80 | |
GenBank mRNA | AK300595.1 | - | 11 - 12 | 1433 | 80 |
AK300605.1 | - | 13 - 14 | 1762 | 80 | |
AK312545.1 | - | 13 - 14 | 1793 | 80 | |
AY513654.1 | - | 10 - 11 | 1324 | 80 | |
AY513655.1 | - | 10 - 11 | 1324 | 80 | |
AY513656.1 | - | 11 - 12 | 1534 | 80 | |
AY513657.1 | - | 11 - 12 | 1498 | 80 | |
BC035357.1 | - | 13 - 14 | 1780 | 80 | |
X52192.1 | - | 13 - 14 | 1783 | 80 |