Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MICAL2
Entrez Gene ID
9645
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 2
Gene Aliases
MICAL-2, MICAL2PV1, MICAL2PV2
Chromosome Location
Chr.11: 12110576 - 12281017 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282663.1 NP_001269592.1 21 - 22 3082 78
NM_014632.3 NP_055447.1 21 - 22 3086 78
XM_011520490.2 XP_011518792.1 19 - 20 2789 78
XM_011520491.2 XP_011518793.1 19 - 20 2789 78
XM_017018589.1 XP_016874078.1 20 - 21 2902 78
XM_017018590.1 XP_016874079.1 20 - 21 2947 78
GenBank mRNA AB018293.1 - 21 - 22 3047 78
BC044577.1 - 21 - 22 3071 78

View Full Product Details