The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288632.1 | NP_001275561.1 | 1 - 2 | 155 | 161 |
NM_001288636.1 | NP_001275565.1 | 1 - 2 | 155 | 263 | |
NM_001288638.1 | NP_001275567.1 | 1 - 2 | 155 | 161 | |
NM_001288639.1 | NP_001275568.1 | 1 - 2 | 155 | 113 | |
NM_018182.3 | NP_060652.2 | 1 - 2 | 155 | 113 | |
GenBank mRNA | AK001562.1 | - | 1 - 2 | 106 | 113 |
DA185693.1 | - | 1 - 2 | 108 | 113 | |
DB453514.1 | - | 1 - 2 | 104 | 161 | |
DC374179.1 | - | 1 - 2 | 85 | 113 | |
DC376581.1 | - | 1 - 2 | 101 | 113 | |
DC399026.1 | - | 1 - 2 | 157 | 161 | |
DC400943.1 | - | 1 - 2 | 69 | 161 | |
HY006495.1 | - | 1 - 2 | 140 | 263 |