The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113182.2 | NP_001106653.1 | 11 - 12 | 3755 | 69 |
NM_001199455.1 | NP_001186384.1 | 11 - 12 | 3550 | 69 | |
NM_001199456.1 | NP_001186385.1 | 10 - 11 | 2158 | 69 | |
NM_001291986.1 | NP_001278915.1 | 12 - 13 | 3847 | 69 | |
NM_005104.3 | NP_005095.1 | 11 - 12 | 3842 | 69 | |
GenBank mRNA | AL832722.1 | - | 12 - 13 | 3782 | 69 |
BC063840.1 | - | 11 - 12 | 2415 | 69 | |
BX647233.1 | - | 10 - 11 | 2158 | 69 | |
BX648109.1 | - | 11 - 12 | 3756 | 69 | |
D42040.1 | - | 11 - 12 | 3842 | 69 | |
M80613.1 | - | 10 - 11 | 3177 | 69 | |
X62083.1 | - | 10 - 11 | 3177 | 69 |