The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193476.1 | NP_001180405.1 | 3 - 4 | 444 | 78 |
NM_052961.3 | NP_443193.1 | 3 - 4 | 468 | 78 | |
NM_138718.2 | NP_619732.2 | 3 - 4 | 506 | 78 | |
XM_011514294.2 | XP_011512596.1 | 3 - 4 | 687 | 78 | |
XM_017010235.1 | XP_016865724.1 | 3 - 4 | 494 | 78 | |
GenBank mRNA | AF314959.1 | - | 2 - 3 | 338 | 78 |
AF331522.1 | - | 3 - 4 | 412 | 78 | |
AK122928.1 | - | 3 - 4 | 444 | 78 | |
AK125189.1 | - | 3 - 4 | 688 | 78 | |
BC025408.2 | - | 3 - 4 | 499 | 78 | |
BG702985.1 | - | 3 - 4 | 78 |