The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000287.3 | NP_000278.3 | 15 - 16 | 2757 | 107 |
NM_001316313.1 | NP_001303242.1 | 15 - 16 | 2493 | 107 | |
NR_133009.1 | 13 - 14 | 2510 | 107 | ||
XM_011514661.1 | XP_011512963.1 | 14 - 15 | 2673 | 107 | |
GenBank mRNA | AB051076.1 | - | 15 - 16 | 2664 | 107 |
AB051077.1 | - | 15 - 16 | 2400 | 107 | |
AB051078.1 | - | 13 - 14 | 2417 | 107 | |
AK314237.1 | - | 15 - 16 | 2693 | 107 | |
BC033487.1 | - | 15 - 16 | 2672 | 107 | |
BC048331.1 | - | 15 - 16 | 2680 | 107 | |
D83703.1 | - | 15 - 16 | 2734 | 107 |