Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX6
Entrez Gene ID
5190
Gene Name
peroxisomal biogenesis factor 6
Gene Aliases
HMLR2, PAF-2, PAF2, PBD4A, PDB4B, PXAAA1
Chromosome Location
Chr.6: 42963873 - 42979243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000287.3 NP_000278.3 7 - 8 1788 77
NM_001316313.1 NP_001303242.1 7 - 8 1524 77
NR_133009.1 7 - 8 1788 77
XM_011514661.1 XP_011512963.1 6 - 7 1704 77
GenBank mRNA AB051076.1 - 7 - 8 1695 77
AB051077.1 - 7 - 8 1431 77
AB051078.1 - 7 - 8 1695 77
AB208906.1 - 7 - 8 1748 77
AK225704.1 - 6 - 7 2101 77
AK314237.1 - 7 - 8 1724 77
BC033487.1 - 7 - 8 1703 77
BC048331.1 - 7 - 8 1711 77
D83703.1 - 7 - 8 1765 77

View Full Product Details