Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTNAP5
Entrez Gene ID
129684
Gene Name
contactin associated protein like 5
Gene Aliases
caspr5
Chromosome Location
Chr.2: 124025287 - 124921201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130773.3 NP_570129.1 16 - 17 2894 98
XM_006712258.1 XP_006712321.1 16 - 17 2897 98
XM_017003316.1 XP_016858805.1 16 - 17 2897 98
GenBank mRNA AB077881.1 - 16 - 17 2530 98
AK056528.1 - 16 - 17 2897 98

View Full Product Details