The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130773.3 | NP_570129.1 | 4 - 5 | 888 | 74 |
XM_006712258.1 | XP_006712321.1 | 4 - 5 | 888 | 74 | |
XM_017003316.1 | XP_016858805.1 | 4 - 5 | 888 | 74 | |
GenBank mRNA | AB077881.1 | - | 4 - 5 | 524 | 74 |
AK056528.1 | - | 4 - 5 | 888 | 74 |