The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001315529.1 | NP_001302458.1 | 13 - 14 | 2421 | 103 |
NM_006031.5 | NP_006022.3 | 13 - 14 | 2259 | 103 | |
XM_005261124.4 | XP_005261181.1 | 13 - 14 | 2278 | 103 | |
XM_011529594.2 | XP_011527896.1 | 14 - 15 | 2359 | 103 | |
XM_017028362.1 | XP_016883851.1 | 13 - 14 | 2278 | 103 | |
XM_017028363.1 | XP_016883852.1 | 13 - 14 | 2447 | 103 | |
GenBank mRNA | AB007862.3 | - | 13 - 14 | 2422 | 103 |
AF515282.1 | - | 13 - 14 | 2152 | 103 | |
BC035913.1 | - | 14 - 15 | 2201 | 103 | |
U52962.1 | - | 13 - 14 | 2204 | 103 |