Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCNT
Entrez Gene ID
5116
Gene Name
pericentrin
Gene Aliases
KEN, MOPD2, PCN, PCNT2, PCNTB, PCTN2, SCKL4
Chromosome Location
Chr.21: 46324103 - 46445769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001315529.1 NP_001302458.1 5 - 6 1244 109
NM_006031.5 NP_006022.3 5 - 6 1082 109
XM_005261124.4 XP_005261181.1 5 - 6 1101 109
XM_011529594.2 XP_011527896.1 6 - 7 1182 109
XM_017028362.1 XP_016883851.1 5 - 6 1101 109
XM_017028363.1 XP_016883852.1 5 - 6 1270 109
GenBank mRNA AB007862.3 - 5 - 6 1245 109
AF515282.1 - 5 - 6 975 109
BC035913.1 - 6 - 7 1024 109
U52962.1 - 5 - 6 1027 109

View Full Product Details