Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCIN
Entrez Gene ID
85477
Gene Name
scinderin
Gene Aliases
-
Chromosome Location
Chr.7: 12570349 - 12653603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112706.2 NP_001106177.1 1 - 2 403 69
GenBank mRNA AB067492.1 - 1 - 2 403 69
AF276507.1 - 1 - 2 193 69
AK075123.1 - 1 - 2 262 69
AK290363.1 - 1 - 2 296 69
BC021090.1 - 1 - 2 69

View Full Product Details