The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172437.2 | NP_001165908.1 | 1 - 2 | 268 | 98 |
NM_001172438.2 | NP_001165909.1 | 1 - 2 | 268 | 98 | |
GenBank mRNA | AK299837.1 | - | - | 223 | 98 |