The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014924.4 | NP_055739.2 | 4 - 5 | 450 | 121 |
XM_011536563.2 | XP_011534865.1 | 2 - 3 | 154 | 121 | |
GenBank mRNA | AK131251.1 | - | 4 - 5 | 450 | 121 |
AK292810.1 | - | 4 - 5 | 422 | 121 | |
BC109089.1 | - | 4 - 5 | 435 | 121 | |
BC109090.1 | - | 4 - 5 | 435 | 121 |