The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320714.1 | NP_001307643.1 | 21 - 22 | 5187 | 110 |
NM_005128.3 | NP_005119.2 | 21 - 22 | 5078 | 110 | |
XM_017028509.1 | XP_016883998.1 | 18 - 19 | 4581 | 110 | |
GenBank mRNA | AB023150.2 | - | 18 - 19 | 4547 | 110 |
AJ237839.2 | - | 21 - 22 | 5084 | 110 | |
AK025095.1 | - | 3 - 4 | 679 | 110 | |
BC012791.2 | - | 3 - 4 | 720 | 110 |