The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320714.1 | NP_001307643.1 | 26 - 27 | 5677 | 67 |
NM_005128.3 | NP_005119.2 | 26 - 27 | 5568 | 67 | |
XM_017028509.1 | XP_016883998.1 | 23 - 24 | 5071 | 67 | |
GenBank mRNA | AB023150.2 | - | 23 - 24 | 5037 | 67 |
AJ237839.2 | - | 26 - 27 | 5574 | 67 | |
AK026478.1 | - | 5 - 6 | 504 | 67 | |
BC012791.2 | - | 8 - 9 | 1210 | 67 |