Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSMD2
Entrez Gene ID
114784
Gene Name
CUB and Sushi multiple domains 2
Gene Aliases
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
Chromosome Location
Chr.1: 33513999 - 34165842 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281956.1 NP_001268885.1 62 - 63 10057 97
NM_052896.4 NP_443128.2 61 - 62 9477 97
XM_017000185.1 XP_016855674.1 62 - 63 9976 97
XM_017000186.1 XP_016855675.1 62 - 63 9976 97
XM_017000187.1 XP_016855676.1 62 - 63 9805 97
XM_017000188.1 XP_016855677.1 62 - 63 9714 97
XM_017000189.1 XP_016855678.1 62 - 63 9976 97
XM_017000190.1 XP_016855679.1 56 - 57 9241 97
XM_017000194.1 XP_016855683.1 62 - 63 9714 97
GenBank mRNA AB067471.1 - 11 - 12 1827 97
AB212622.1 - 62 - 63 10057 97
AK095627.1 - 4 - 5 544 97
AY210418.1 - 61 - 62 9477 97

View Full Product Details