Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNF362
Entrez Gene ID
149076
Gene Name
zinc finger protein 362
Gene Aliases
RN, lin-29
Chromosome Location
Chr.1: 33256307 - 33300719 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152493.2 NP_689706.2 7 - 8 1158 75
XM_005270507.2 XP_005270564.1 6 - 7 1298 75
XM_005270508.4 XP_005270565.1 7 - 8 1955 75
XM_017000415.1 XP_016855904.1 6 - 7 1870 75
GenBank mRNA AF318356.1 - 6 - 7 1008 75
BC104779.1 - 6 - 7 1080 75
BC112113.1 - 6 - 7 1080 75

View Full Product Details