The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320724.1 | NP_001307653.1 | 3 - 4 | 493 | 155 |
NM_016940.2 | NP_058636.1 | 2 - 3 | 388 | 155 | |
GenBank mRNA | AF212232.1 | - | 2 - 3 | 349 | 155 |
AK000056.1 | - | 2 - 3 | 392 | 155 | |
AK223357.1 | - | 2 - 3 | 399 | 155 | |
BC012546.1 | - | 2 - 3 | 356 | 155 | |
BC017912.1 | - | 2 - 3 | 374 | 155 | |
DB039085.1 | - | 3 - 4 | 490 | 155 | |
Y19009.1 | - | 2 - 3 | 367 | 155 |