The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202469.1 | NP_001189398.1 | 1 - 2 | 138 | |
NM_001483.2 | NP_001474.1 | 1 - 2 | 138 | ||
GenBank mRNA | AK097049.1 | - | 1 - 2 | 138 | |
AK315648.1 | - | 1 - 2 | 109 | 137 | |
BC030821.1 | - | 1 - 2 | 138 | ||
CX166452.1 | - | 1 - 2 | 138 |