Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMA2
Entrez Gene ID
3908
Gene Name
laminin subunit alpha 2
Gene Aliases
LAMM
Chromosome Location
Chr.6: 128883141 - 129516566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000426.3 NP_000417.2 48 - 49 6974 74
NM_001079823.1 NP_001073291.1 48 - 49 6974 74
XM_005266981.3 XP_005267038.1 49 - 50 7238 74
XM_005266982.3 XP_005267039.1 49 - 50 7238 74
XM_011535820.2 XP_011534122.1 48 - 49 7232 74
XM_017010851.1 XP_016866340.1 49 - 50 9626 74
XM_017010852.1 XP_016866341.1 38 - 39 8515 74
GenBank mRNA AB208922.1 - 23 - 24 3076 74
M59832.1 - 8 - 9 929 74
Z26653.1 - 48 - 49 6918 74

View Full Product Details