Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMA2
Entrez Gene ID
3908
Gene Name
laminin subunit alpha 2
Gene Aliases
LAMM
Chromosome Location
Chr.6: 128883141 - 129516566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000426.3 NP_000417.2 63 - 64 9099 147
NM_001079823.1 NP_001073291.1 62 - 63 9087 147
XM_005266981.3 XP_005267038.1 64 - 65 9363 147
XM_005266982.3 XP_005267039.1 63 - 64 9351 147
XM_011535820.2 XP_011534122.1 63 - 64 9357 147
XM_017010851.1 XP_016866340.1 64 - 65 11751 147
XM_017010852.1 XP_016866341.1 53 - 54 10640 147
GenBank mRNA AB208922.1 - 37 - 38 5189 147
M59832.1 - 23 - 24 3054 147

View Full Product Details