The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004311.3 | NP_004302.1 | 2 - 3 | 303 | 109 |
XM_017016260.1 | XP_016871749.1 | 2 - 3 | 489 | 109 | |
GenBank mRNA | AF493889.1 | - | 1 - 2 | 145 | 109 |
AK312525.1 | - | 2 - 3 | 255 | 109 | |
BC009841.2 | - | 2 - 3 | 200 | 109 | |
CR407637.1 | - | 1 - 2 | 145 | 109 | |
U07151.1 | - | 1 - 2 | 160 | 109 |