The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184740.1 | NP_001171669.1 | 9 - 10 | 891 | 73 |
NM_005231.3 | NP_005222.2 | 9 - 10 | 891 | 73 | |
NM_138565.2 | NP_612632.1 | 9 - 10 | 891 | 73 | |
XM_006718448.3 | XP_006718511.1 | 9 - 10 | 891 | 73 | |
XM_017017312.1 | XP_016872801.1 | 9 - 10 | 891 | 73 | |
GenBank mRNA | AK222613.1 | - | 9 - 10 | 860 | 73 |
AK291097.1 | - | 9 - 10 | 862 | 73 | |
BC008799.2 | - | 9 - 10 | 853 | 73 | |
BC033889.1 | - | 9 - 10 | 861 | 73 | |
M98343.1 | - | 9 - 10 | 853 | 73 |