The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193288.2 | NP_001180217.1 | 6 - 7 | 1579 | 91 |
NM_020344.3 | NP_065077.1 | 7 - 8 | 1630 | 91 | |
XM_005251425.2 | XP_005251482.1 | 8 - 9 | 1909 | 91 | |
XM_005251426.4 | XP_005251483.1 | 8 - 9 | 2441 | 91 | |
XM_006716750.3 | XP_006716813.1 | 7 - 8 | 2390 | 91 | |
XM_017014592.1 | XP_016870081.1 | 12 - 13 | 2380 | 91 | |
XM_017014593.1 | XP_016870082.1 | 7 - 8 | 1862 | 91 | |
GenBank mRNA | AF097366.1 | - | 7 - 8 | 1539 | 91 |
AF177987.1 | - | 6 - 7 | 1488 | 91 | |
BC069622.1 | - | 7 - 8 | 1527 | 91 | |
BC143889.1 | - | 6 - 7 | 1449 | 91 |