The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193288.2 | NP_001180217.1 | 8 - 9 | 1833 | 85 |
NM_020344.3 | NP_065077.1 | 9 - 10 | 1884 | 85 | |
XM_005251425.2 | XP_005251482.1 | 10 - 11 | 2163 | 85 | |
XM_005251426.4 | XP_005251483.1 | 10 - 11 | 2695 | 85 | |
XM_006716750.3 | XP_006716813.1 | 9 - 10 | 2644 | 85 | |
XM_017014592.1 | XP_016870081.1 | 14 - 15 | 2634 | 85 | |
XM_017014593.1 | XP_016870082.1 | 9 - 10 | 2116 | 85 | |
GenBank mRNA | AF097366.1 | - | 9 - 10 | 1793 | 85 |
AF177987.1 | - | 8 - 9 | 1742 | 85 | |
BC069622.1 | - | 9 - 10 | 1781 | 85 | |
BC143889.1 | - | 8 - 9 | 1703 | 85 |