The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104546.1 | NP_001098016.1 | 4 - 5 | 308 | 115 |
NM_006413.4 | NP_006404.1 | 4 - 5 | 308 | 115 | |
XM_011539120.2 | XP_011537422.1 | 7 - 8 | 861 | 115 | |
XM_011539121.2 | XP_011537423.1 | 7 - 8 | 861 | 115 | |
XM_017015475.1 | XP_016870964.1 | 7 - 8 | 861 | 115 | |
XM_017015476.1 | XP_016870965.1 | 7 - 8 | 861 | 115 | |
XM_017015477.1 | XP_016870966.1 | 5 - 6 | 561 | 115 | |
XM_017015478.1 | XP_016870967.1 | 7 - 8 | 861 | 115 | |
GenBank mRNA | AK296196.1 | - | 4 - 5 | 286 | 115 |
AK312900.1 | - | 4 - 5 | 282 | 115 | |
AL833628.1 | - | 4 - 5 | 294 | 115 | |
BC006991.1 | - | 5 - 6 | 567 | 115 | |
DB049454.1 | - | 4 - 5 | 308 | 115 | |
U77665.1 | - | 4 - 5 | 300 | 115 |