The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001560.2 | NP_001551.1 | 8 - 9 | 1050 | 89 |
XM_011531336.1 | XP_011529638.1 | 8 - 9 | 1328 | 89 | |
XM_017029507.1 | XP_016884996.1 | 7 - 8 | 1067 | 89 | |
GenBank mRNA | AB209849.1 | - | 8 - 9 | 1005 | 89 |
AK296958.1 | - | 3 - 4 | 279 | 89 | |
AK313467.1 | - | 8 - 9 | 1039 | 89 | |
BC009960.2 | - | 8 - 9 | 1037 | 89 | |
BC015768.1 | - | 8 - 9 | 1054 | 89 | |
U81379.3 | - | 8 - 9 | 1047 | 89 | |
Y09328.1 | - | 8 - 9 | 1040 | 89 | |
Y10659.1 | - | 8 - 9 | 1050 | 89 |