The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031442.3 | NP_113630.1 | 1 - 2 | 489 | 77 |
GenBank mRNA | AL136550.1 | - | 1 - 2 | 239 | 77 |
BC039242.1 | - | 1 - 2 | 489 | 77 | |
CX788638.1 | - | 1 - 2 | 477 | 77 |