The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001369.2 | NP_001360.1 | 33 - 34 | 5528 | 99 |
XM_005248262.3 | XP_005248319.2 | 33 - 34 | 5611 | 99 | |
XM_017009177.1 | XP_016864666.1 | 33 - 34 | 5611 | 99 | |
XM_017009178.1 | XP_016864667.1 | 33 - 34 | 6414 | 99 | |
XM_017009179.1 | XP_016864668.1 | 27 - 28 | 4708 | 99 | |
XM_017009180.1 | XP_016864669.1 | 33 - 34 | 5611 | 99 | |
XM_017009181.1 | XP_016864670.1 | 33 - 34 | 5611 | 99 | |
XM_017009182.1 | XP_016864671.1 | 33 - 34 | 5611 | 99 | |
XM_017009183.1 | XP_016864672.1 | 33 - 34 | 5611 | 99 | |
XM_017009184.1 | XP_016864673.1 | 33 - 34 | 5611 | 99 | |
XM_017009185.1 | XP_016864674.1 | 5 - 6 | 907 | 99 | |
XM_017009186.1 | XP_016864675.1 | 2 - 3 | 323 | 99 | |
XM_017009187.1 | XP_016864676.1 | 33 - 34 | 5611 | 99 | |
GenBank mRNA | AY045575.1 | - | 33 - 34 | 5528 | 99 |