The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256946.1 | NP_001243875.1 | 2 - 3 | 224 | 88 |
NM_001282670.1 | NP_001269599.1 | 6 - 7 | 3070 | 88 | |
NM_001282671.1 | NP_001269600.1 | 2 - 3 | 224 | 88 | |
NM_001282672.1 | NP_001269601.1 | 2 - 3 | 224 | 88 | |
NM_001282673.1 | NP_001269602.1 | 2 - 3 | 224 | 88 | |
NM_182533.2 | NP_872339.2 | 2 - 3 | 224 | 88 | |
XM_006710419.3 | XP_006710482.1 | 3 - 4 | 809 | 88 | |
XM_011540914.2 | XP_011539216.1 | 7 - 8 | 760 | 88 | |
XM_011540921.2 | XP_011539223.1 | 4 - 5 | 609 | 88 | |
XM_011540922.1 | XP_011539224.1 | 2 - 3 | 223 | 88 | |
XM_017000553.1 | XP_016856042.1 | 5 - 6 | 607 | 88 | |
GenBank mRNA | AF461903.1 | - | 2 - 3 | 2038 | 88 |
AF469129.1 | - | 2 - 3 | 190 | 88 | |
AK055593.1 | - | 2 - 3 | 224 | 88 | |
AK126870.1 | - | 6 - 7 | 3069 | 88 | |
BC041041.1 | - | 2 - 3 | 163 | 88 | |
BC066360.1 | - | 2 - 3 | 221 | 88 | |
BC103992.2 | - | 2 - 3 | 224 | 88 | |
BG825658.1 | - | 2 - 3 | 205 | 89 | |
BU542153.1 | - | 1 - 2 | 49 | 88 | |
BU626113.1 | - | 2 - 3 | 88 | ||
DA596011.1 | - | 2 - 3 | 224 | 88 |