Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- YBX3
- Entrez Gene ID
-
8531
- Gene Name
- Y-box binding protein 3
- Gene Aliases
-
CSDA, CSDA1, DBPA, ZONAB
- Chromosome Location
-
Chr.12: 10699077 - 10723448 on Build GRCh38
- Gene Symbol
- YBX3P1
- Entrez Gene ID
-
440359
- Gene Name
- Y-box binding protein 3 pseudogene 1
- Gene Aliases
-
CSDAP1
- Chromosome Location
-
Chr.16: 31567767 - 31569524 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027011.1 |
|
1 - 1 |
291 |
69 |
View Full Product Details