The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145426.1 | NP_001138898.1 | 8 - 9 | 1188 | 88 |
NM_003651.4 | NP_003642.3 | 9 - 10 | 1395 | 88 | |
XM_017020122.1 | XP_016875611.1 | 9 - 10 | 1480 | 88 | |
XM_017020123.1 | XP_016875612.1 | 8 - 9 | 1273 | 88 | |
GenBank mRNA | AB209896.1 | - | 7 - 8 | 5965 | 88 |
BC008801.2 | - | 9 - 10 | 1353 | 88 | |
BC009744.2 | - | 8 - 9 | 1141 | 88 | |
BC015564.1 | - | 9 - 10 | 1344 | 88 | |
BC015913.1 | - | 8 - 9 | 1130 | 88 | |
BC021926.1 | - | 9 - 10 | 1346 | 88 | |
X72712.1 | - | 9 - 10 | 1373 | 88 |