Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTLN
Entrez Gene ID
54875
Gene Name
centlein
Gene Aliases
C9orf101, C9orf39, bA340N12.1
Chromosome Location
Chr.9: 17134991 - 17510019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017738.3 NP_060208.2 12 - 13 1965 128
XM_005251492.1 XP_005251549.1 12 - 13 1965 128
XM_006716793.3 XP_006716856.1 12 - 13 1763 128
XM_011517941.2 XP_011516243.1 12 - 13 1965 128
XM_017014839.1 XP_016870328.1 12 - 13 1965 128
XM_017014840.1 XP_016870329.1 12 - 13 1965 128
XM_017014841.1 XP_016870330.1 11 - 12 1770 128
XM_017014842.1 XP_016870331.1 12 - 13 1965 128
XM_017014843.1 XP_016870332.1 12 - 13 1965 128
XM_017014844.1 XP_016870333.1 5 - 6 641 128
XM_017014845.1 XP_016870334.1 3 - 4 1446 128
GenBank mRNA AK000283.1 - 8 - 9 1060 128
AK308837.1 - 12 - 13 1920 128
BX647069.1 - 12 - 13 1924 128

View Full Product Details