Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTLN
Entrez Gene ID
54875
Gene Name
centlein
Gene Aliases
C9orf101, C9orf39, bA340N12.1
Chromosome Location
Chr.9: 17134991 - 17510019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114395.2 NP_001107867.1 1 - 2 438 72
NM_001286984.1 NP_001273913.1 1 - 2 438 72
NM_017738.3 NP_060208.2 1 - 2 438 72
XM_005251492.1 XP_005251549.1 1 - 2 438 72
XM_011517941.2 XP_011516243.1 1 - 2 438 72
XM_017014839.1 XP_016870328.1 1 - 2 438 72
XM_017014840.1 XP_016870329.1 1 - 2 438 72
XM_017014841.1 XP_016870330.1 1 - 2 438 72
XM_017014842.1 XP_016870331.1 1 - 2 438 72
XM_017014843.1 XP_016870332.1 1 - 2 438 72
GenBank mRNA AK098502.1 - 1 - 2 389 72
AK303973.1 - 1 - 2 396 72
AK308837.1 - 1 - 2 393 72
BX647069.1 - 1 - 2 396 72
DA482759.1 - 1 - 2 390 72
DB079230.1 - 1 - 2 439 72

View Full Product Details