The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017738.3 | NP_060208.2 | 24 - 25 | 4111 | 113 |
XM_005251492.1 | XP_005251549.1 | 24 - 25 | 4108 | 113 | |
XM_006716793.3 | XP_006716856.1 | 24 - 25 | 3909 | 113 | |
XM_017014839.1 | XP_016870328.1 | 24 - 25 | 4111 | 113 | |
XM_017014840.1 | XP_016870329.1 | 24 - 25 | 4111 | 113 | |
XM_017014841.1 | XP_016870330.1 | 23 - 24 | 3916 | 113 | |
XM_017014844.1 | XP_016870333.1 | 17 - 18 | 2787 | 113 | |
XM_017014845.1 | XP_016870334.1 | 15 - 16 | 3592 | 113 | |
XM_017014846.1 | XP_016870335.1 | 12 - 13 | 2115 | 113 | |
XM_017014847.1 | XP_016870336.1 | 12 - 13 | 2108 | 113 | |
GenBank mRNA | AK000283.1 | - | 20 - 21 | 3206 | 113 |
AK308837.1 | - | 24 - 25 | 4063 | 113 |