Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTLN
Entrez Gene ID
54875
Gene Name
centlein
Gene Aliases
C9orf101, C9orf39, bA340N12.1
Chromosome Location
Chr.9: 17134991 - 17510019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114395.2 NP_001107867.1 3 - 4 605 91
NM_017738.3 NP_060208.2 3 - 4 605 91
XM_005251492.1 XP_005251549.1 3 - 4 605 91
XM_006716793.3 XP_006716856.1 3 - 4 403 91
XM_011517941.2 XP_011516243.1 3 - 4 605 91
XM_017014839.1 XP_016870328.1 3 - 4 605 91
XM_017014840.1 XP_016870329.1 3 - 4 605 91
XM_017014841.1 XP_016870330.1 3 - 4 605 91
XM_017014842.1 XP_016870331.1 3 - 4 605 91
XM_017014843.1 XP_016870332.1 3 - 4 605 91
GenBank mRNA AK098502.1 - 3 - 4 556 91
AK308837.1 - 3 - 4 560 91
BX647069.1 - 3 - 4 563 91

View Full Product Details