The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297614.2 | NP_001284543.1 | 13 - 14 | 1706 | 160 |
NM_001304479.1 | NP_001291408.1 | 13 - 14 | 1451 | 160 | |
NM_020468.5 | NP_065201.1 | 11 - 12 | 1574 | 160 | |
NM_153816.5 | NP_722523.1 | 13 - 14 | 1706 | 160 | |
XM_005248738.3 | XP_005248795.1 | 13 - 14 | 1679 | 160 | |
XM_005248740.3 | XP_005248797.1 | 11 - 12 | 1548 | 160 | |
XM_011535977.2 | XP_011534279.1 | 12 - 13 | 1122 | 160 | |
XM_011535979.1 | XP_011534281.1 | 11 - 12 | 1114 | 160 | |
XM_017011090.1 | XP_016866579.1 | 13 - 14 | 1188 | 160 | |
XM_017011091.1 | XP_016866580.1 | 13 - 14 | 1679 | 160 | |
XM_017011092.1 | XP_016866581.1 | 11 - 12 | 1316 | 160 | |
XM_017011093.1 | XP_016866582.1 | 13 - 14 | 1393 | 160 | |
XM_017011094.1 | XP_016866583.1 | 11 - 12 | 1288 | 160 | |
XM_017011095.1 | XP_016866584.1 | 11 - 12 | 1028 | 160 | |
XM_017011096.1 | XP_016866585.1 | 13 - 14 | 1417 | 160 | |
XM_017011097.1 | XP_016866586.1 | 13 - 14 | 1401 | 160 | |
XM_017011098.1 | XP_016866587.1 | 12 - 13 | 1319 | 160 | |
XM_017011099.1 | XP_016866588.1 | 11 - 12 | 1285 | 160 | |
XM_017011100.1 | XP_016866589.1 | 10 - 11 | 1009 | 160 | |
XM_017011101.1 | XP_016866590.1 | 12 - 13 | 1118 | 160 | |
GenBank mRNA | AK000362.1 | - | 13 - 14 | 160 | |
AK026479.1 | - | 12 - 13 | 1838 | 160 | |
AK095380.1 | - | 14 - 15 | 1901 | 160 | |
AK295417.1 | - | 13 - 14 | 1180 | 160 | |
AY044865.1 | - | 11 - 12 | 1115 | 160 | |
BC005110.1 | - | 11 - 12 | 1303 | 160 | |
BC046520.2 | - | 13 - 14 | 1161 | 160 | |
BC095419.1 | - | 13 - 14 | 1291 | 160 |