Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX14
Entrez Gene ID
57231
Gene Name
sorting nexin 14
Gene Aliases
RGS-PX2, SCAR20
Chromosome Location
Chr.6: 85505496 - 85594156 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297614.2 NP_001284543.1 8 - 9 1225 146
NM_001304479.1 NP_001291408.1 8 - 9 970 146
NM_020468.5 NP_065201.1 6 - 7 1093 146
NM_153816.5 NP_722523.1 8 - 9 1225 146
XM_005248738.3 XP_005248795.1 8 - 9 1198 146
XM_005248740.3 XP_005248797.1 6 - 7 1067 146
XM_011535977.2 XP_011534279.1 7 - 8 641 146
XM_011535979.1 XP_011534281.1 6 - 7 633 146
XM_017011090.1 XP_016866579.1 8 - 9 707 146
XM_017011091.1 XP_016866580.1 8 - 9 1198 146
XM_017011092.1 XP_016866581.1 6 - 7 835 146
XM_017011093.1 XP_016866582.1 8 - 9 912 146
XM_017011094.1 XP_016866583.1 6 - 7 807 146
XM_017011095.1 XP_016866584.1 6 - 7 547 146
XM_017011096.1 XP_016866585.1 8 - 9 940 146
XM_017011097.1 XP_016866586.1 8 - 9 892 146
XM_017011098.1 XP_016866587.1 7 - 8 842 146
XM_017011099.1 XP_016866588.1 6 - 7 808 146
XM_017011100.1 XP_016866589.1 5 - 6 500 146
XM_017011101.1 XP_016866590.1 7 - 8 641 146
GenBank mRNA AK026479.1 - 7 - 8 1357 146
AK095380.1 - 9 - 10 1424 146
AK125063.1 - 4 - 5 1277 146
AK295417.1 - 8 - 9 699 146
AY044865.1 - 6 - 7 634 146
BC005110.1 - 6 - 7 822 146
BC046520.2 - 8 - 9 680 146
BC068589.1 - 8 - 9 964 146
BC095419.1 - 8 - 9 810 146

View Full Product Details