The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291702.1 | NP_001278631.1 | 1 - 2 | 284 | 154 |
NM_001291703.1 | NP_001278632.1 | 1 - 2 | 284 | 154 | |
GenBank mRNA | AB435052.1 | - | 1 - 2 | 211 | 154 |
AB435053.1 | - | 1 - 2 | 211 | 154 | |
AB435054.1 | - | 1 - 2 | 211 | 154 | |
AB435055.1 | - | 1 - 2 | 211 | 154 | |
AB435056.1 | - | 1 - 2 | 211 | 154 | |
AB435057.1 | - | 1 - 2 | 211 | 154 | |
AB435058.1 | - | 1 - 2 | 211 | 154 | |
AB435059.1 | - | 1 - 2 | 211 | 154 | |
AB435061.1 | - | 1 - 2 | 211 | 154 | |
AJ238982.1 | - | 1 - 2 | 256 | 154 | |
AK075110.1 | - | 1 - 2 | 283 | 154 | |
AK314708.1 | - | 1 - 2 | 284 | 154 | |
BC104416.2 | - | 1 - 2 | 271 | 154 | |
BC104417.1 | - | 1 - 2 | 271 | 154 | |
BC113022.1 | - | 1 - 2 | 271 | 154 | |
BC113023.1 | - | 1 - 2 | 271 | 154 | |
BC148317.2 | - | 1 - 2 | 271 | 154 | |
DA670661.1 | - | 1 - 2 | 285 | 154 |