Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VNN3
Entrez Gene ID
55350
Gene Name
vanin 3
Gene Aliases
HSA238982
Chromosome Location
Chr.6: 132722787 - 132734765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291703.1 NP_001278632.1 2 - 3 413 63
GenBank mRNA AB435055.1 - 2 - 3 340 63
AB435057.1 - 2 - 3 340 63
AJ238982.1 - 2 - 3 385 63
BC113022.1 - 2 - 3 400 63
BC113023.1 - 2 - 3 400 63
BC148317.2 - 2 - 3 400 63

View Full Product Details