The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300913.1 | NP_001287842.1 | 3 - 4 | 521 | 70 |
NM_022338.3 | NP_071733.1 | 3 - 4 | 521 | 70 | |
XM_005274053.3 | XP_005274110.1 | 3 - 4 | 696 | 70 | |
GenBank mRNA | AF264781.1 | - | 3 - 4 | 481 | 70 |
AF370372.1 | - | 2 - 3 | 70 | ||
AY358754.1 | - | 1 - 2 | 83 | 70 | |
BC011765.2 | - | 2 - 3 | 370 | 70 | |
BG701401.1 | - | 3 - 4 | 526 | 70 | |
BI862360.1 | - | 3 - 4 | 495 | 70 | |
BX384793.2 | - | 2 - 3 | 323 | 70 |