Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSG1
Entrez Gene ID
83445
Gene Name
germ cell associated 1
Gene Aliases
-
Chromosome Location
Chr.12: 13083537 - 13103720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080554.2 NP_001074023.1 5 - 6 942 64
NM_001080555.2 NP_001074024.1 5 - 6 818 64
NM_001206842.1 NP_001193771.1 4 - 5 749 64
NM_031289.3 NP_112579.2 4 - 5 849 64
NM_153823.3 NP_722545.2 4 - 5 725 64
XM_005253493.2 XP_005253550.1 5 - 6 872 64
XM_005253495.1 XP_005253552.1 5 - 6 996 64
GenBank mRNA AB593019.1 - 4 - 5 721 64
AB593020.1 - 4 - 5 597 64
AK058045.1 - 5 - 6 891 64
AK075288.1 - 4 - 5 719 64
AK075322.1 - 4 - 5 595 64
AY358513.1 - 4 - 5 703 64
BC001796.1 - 4 - 5 795 64
BC033854.1 - 4 - 5 692 64

View Full Product Details