The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014939.3 | NP_055754.2 | 25 - 26 | 4087 | 117 |
XM_005258233.2 | XP_005258290.1 | 25 - 26 | 4180 | 117 | |
XM_005258234.2 | XP_005258291.1 | 25 - 26 | 4180 | 117 | |
XM_005258235.3 | XP_005258292.1 | 20 - 21 | 2961 | 117 | |
XM_006722420.3 | XP_006722483.1 | 25 - 26 | 4180 | 117 | |
XM_011525884.2 | XP_011524186.1 | 25 - 26 | 4180 | 117 | |
XM_017025615.1 | XP_016881104.1 | 25 - 26 | 3679 | 117 | |
XM_017025616.1 | XP_016881105.1 | 25 - 26 | 4177 | 117 | |
XM_017025617.1 | XP_016881106.1 | 25 - 26 | 4177 | 117 | |
GenBank mRNA | AB023229.1 | - | 25 - 26 | 3780 | 117 |
AK001662.1 | - | 17 - 18 | 2446 | 117 | |
AL136749.1 | - | 26 - 27 | 3612 | 117 | |
BC127109.1 | - | 25 - 26 | 3641 | 117 | |
BC127110.1 | - | 25 - 26 | 3641 | 117 |